PRECISION™ Specimen Containers, Covidien
Supplier: Cardinal Health
17400
14219-084CS 705.39 USD
14219-084 14219-106
PRECISION™ Specimen Containers, Covidien
Sample Containers General Purpose Containers
Containers for specimen and tissue collection protect against leakage and have clearly marked graduations in ounces and milliliters
Container 14219-084 features an attached ID label. Containers 33000-960 and 14219-084 are ETO sterilized.