Monoject™ ANGEL WING® Blood Collection Sets, Covidien
Supplier: Cardinal Health
8881225190 8881225174 8881225281 8881225299 8881225390 8881225315 8881225273 8881225182 8881225208 8881225265 8881225307 8881225707
14219-054CS 166.45 USD
14219-054 14219-050 14219-212 14219-214 82013-182 14219-064 14219-210 14219-052 14219-056 14219-208 37001-292 84009-740
Monoject™ ANGEL WING® Blood Collection Sets, Covidien
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
All sets feature easy-to-handle textured wings and one-handed activation of the safety mechanism
The blood collection sets with attached blood tube/needle holders feature a unique self-sheathing needle system and superior needle sharpness.
All sets come with a 1.9cm (3/4") needle. Sets with a female luer connector are compatible with all blood drawing equipment. A luer adapter is also available for multi-sample collection.