System 250 Stereo Zoom Microscopess, LX Microscopes by Unitron
Supplier: LX Microscopes by UNITRON
18714
14218-514EA 2666.83 USD
14218-514
System 250 Stereo Zoom Microscopess, LX Microscopes by Unitron
Microscopes
The System 250 comes standard with a binocular stereo zoom microscope and two 10x eyepieces
Additional standard components include boom stand with base and two eyepiece shields that enables easy attachment of either a fluorescent ring light or fiber optic ring light.
Ordering information: Additional eyepieces, eyepiece shields, and lenses sold separately.