Paraffin wax blended with synthetic polymers, Solidification point: 56-57°C (132.8-134.6°F), pellets, Tissue-Tek® VIP® processing/embedding medium
Supplier: SAKURA FINETEK USA INC
|
Warning
|
VIP® Paraffin's unique composition allows for reduced compression sections as thin as 2 µm.
- Low melting point of 56°C protects samples from heat damage
- Will not discolor under normal processing conditions
Leaves no plasticizer residue to clog paraffin lines in tissue processors and embedding centers. Supplied in small pellets for rapid melting.
CAS Number:
8002-74-2 |
Specification Test Results
Medium Composition | Purified paraffin and synthetic resin blend |
Applications | Processing/Embedding |
Additives | None |
Melting Point | 56° to 58°C / 132° to 136°F |
High Temperature Stability | 65°C |
Solid Product Range | Pellets |
Sectioning Range | Down to 2µ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...