VWR® Heavy-Duty Equipment Clamps
Supplier: VWR International
14216-208
14216-208EA 238.34 USD
14216-208
VWR® Heavy-Duty Equipment Clamps
Clamps General Purpose Clamps
These heavy-duty clamps support overhead mixers and homogenizers.
  • Strong aluminum alloy construction
  • Long surface length
  • Easy-to-grip knobs
They fit rods from 6 to 24 mm in diameter. Large, easy-to-grip knobs allow extra tightening. Clamps are made of strong aluminum alloy with brushed finish. Long surface length supports rods to avoid vibration and wobble.