FINE-L-KOTE™ Conformal Coatings, Techspray™
Supplier: ITW Chemtronics
2102-12S
14216-084CS 90.07 USD
14216-084 14216-088
FINE-L-KOTE™ Conformal Coatings, Techspray™
Sprays
A full line of coatings designed to meet various production and individual needs.
  • Formulated to ensure coating success in a variety of environmental situations
  • All FINE-L-KOTE™ coatings are durable, resist moisture and fungus, and contain no CFCs
FINE-L-KOTE™ SR is a chemical-resistant silicone-resin conformal coating durable enough to stand up to harsh environments. FINE-L-KOTE™ UR has a clear and repairable urethane coating that is highly resistant to most solvents and molds. It is also recommended for harsh environments.
All FINE-L-KOTE™ coatings are supplied in a 340g (12oz.) aerosol can.
FINE-L-KOTE™ SR is UL listed.