VORTEX™ 360° Duster, Techspray™
Supplier: ITW Chemtronics
1697-8S
14216-082CS 696.47 USD
14216-082
VORTEX™ 360° Duster, Techspray™
Sprays
Multi-directional, high-volume, high-velocity duster blasts contamination from surfaces, quickly dislodging dust and particles from hard-to-reach areas
Pure, moisture-free inert gas (100% HFC-134a) leaves no residue. Nonflammable. Safe on plastics.