Envi-Ro-Tech™ Antistatic Freeze Sprays, Techspray™
Supplier: ITW Chemtronics
1672-10S
14216-058CS 893.47 USD
14216-058
Envi-Ro-Tech™ Antistatic Freeze Sprays, Techspray™
Freezing Sprays
Made of HFC-134a
  • Quick evaporation
  • Non-ozone depleting
  • Will not contaminate substrate
  • Nonflammable
  • No residue
  • No CFC/HCFCs
Use in static-sensitive areas to troubleshoot intermittently faulting capacitors, resistors, semiconductors, and other defective components. The –40°C (–40°F) freeze spray also detects cold solder joints and oxidized junctions.