Hardened Steel Mortar, Retsch®
Supplier: Retsch
024600018
14213-042EA 2584.76 USD
14213-042
Hardened Steel Mortar, Retsch®
Mortars and Pestles Mortars
Designed for use with Retsch® RM200 Mortar Grinder, this hardened steel mortar is intended for use in less demanding applications, or under rough conditions
Material composition is approximately 86% Fe, 11.5% Cr, and trace amounts of C, Mn, Si, P, and S. Density is approximately 7700kg/m3.