SP Wilmad-LabGlass Soap Film Flowmeters, SP Industries
Supplier: SP Industries
LG-8612-095
14202-464EA 334.69 USD
14202-464 14202-634 14209-012 14210-248
SP Wilmad-LabGlass Soap Film Flowmeters, SP Industries
Flow Meters
Made of borosilicate glass, these soap film bubble flowmeters are used in routine analytical laboratory gas chromatography to measure flow rate of the carrier gas.
A soap bubble, formed in the bottom of the graduated tube, is pushed upward by gas from exit end of the chromatograph. Flow rate is determined by measurement of the volume through which the bubble travels in a defined period of time. Includes LG-4340-100 rubber bulb.