SP Wilmad-LabGlass Allihn Condensers, Soxhlet, SP Industries
Supplier: SP Industries
LG-6900-110
14206-558EA 301.79 USD
14206-558 14208-338 14209-024 14209-840
SP Wilmad-LabGlass Allihn Condensers, Soxhlet, SP Industries
Condensers
Allihn condensers are designed for use within a Soxhlet extraction apparatus.