SP Wilmad-LabGlass Buchner Filter Funnels with Fritted Disc, SP Industries
Supplier: SP Industries
LG-7080-144
14202-328EA 85.51 USD
14202-328 14202-330 14202-332 14205-032 14205-034 14205-290 14205-292 14205-364 14205-366 14205-794 14207-062 14207-064 14207-066 14208-070 14208-156 14208-736 14208-738 14208-740 14209-326 14209-328 14209-330 14210-462 14210-464 14210-466
SP Wilmad-LabGlass Buchner Filter Funnels with Fritted Disc, SP Industries
Funnels Filtration Funnels Büchner Funnels
Used in applications where filter paper would be attacked by the material being filtered, these borosilicate glass Buchner filter funnels with sealed-in fritted glass discs are available in various capacities with Coarse(C), Medium(M), or Fine(F) porosities.
Ordering information: Larger sizes available upon request.