SP Wilmad-LabGlass Reduced Pressure Distilling Apparatus, SP Industries
Supplier: SP Industries
ML-560-700
14203-838EA 300.08 USD
14203-838 14204-212 14208-924
SP Wilmad-LabGlass Reduced Pressure Distilling Apparatus, SP Industries
Distillation Components Distillation Sets
Distilling apparatus includes a three-way bent adapter for a Standard Taper 10/18 thermometer with a 25 mm immersion, a 110 mm Liebig condenser, a 105° vacuum adapter (ML-390-700), a 25 mL Erlenmeyer flask (ML-1230-706), and a pear-shaped 50 mL distilling flask (ML-1160-718).
  • Borosilicate glass
All joints (not including thermometer joint) are a Standard Taper 14/20.