SP Wilmad-LabGlass Offset Adapters, SP Industries
Supplier: SP Industries
LG-1580-100
14203-238EA 113.01 USD
14203-238 14208-850
SP Wilmad-LabGlass Offset Adapters, SP Industries
Adapters Adapters, Conical Ground Joints
These offset adapters are manufactured from borosilicate glass.
  • [ST] outer joint on one end
  • [ST] inner joint on the other end
  • Both joints are same size
Adapters have a Standard Taper outer joint on one end and the same size Standard Taper inner joint on the other.