SP Wilmad-LabGlass Dreschel Gas Washing Bottles, SP Industries
Supplier: SP Industries
LG-3690-100
14202-294EA 87.76 USD
14202-294 14205-276 14206-384 14206-450 14208-824 14210-626
SP Wilmad-LabGlass Dreschel Gas Washing Bottles, SP Industries
Bottles Gas Washing Bottles
These Dreschel tall form gas washing bottles are made of borosilicate glass and include a stopper component that contains gas inlet and outlet tubes.
Bottle and stopper are joined by a full-length standard taper 29/42 joint. Complete units are supplied with a plastic clamp (LG-1004-110).