SP Wilmad-LabGlass Heavy-Duty Glass Stirrer Shafts, SP Industries
Supplier: SP Industries
LG-9504-108
14208-392EA 284.31 USD
14208-392 14208-394 14209-560 14209-562
SP Wilmad-LabGlass Heavy-Duty Glass Stirrer Shafts, SP Industries
Stirrers Stirring Shafts
Made of borosilicate glass, these heavy-duty, precision ground, polished stirrer shafts have a 19 mm diameter and are equipped with a side button for attaching stirrer blades (LG-9515 or LG-9516T).
The rod length below contact surface of bearing is unground and varies with flask capacity.