SP Wilmad-LabGlass Specific Gravity/Pycnometer Bottle Apparatus, SP Industries
Supplier: SP Industries
LG-3550-104
14205-834EA 391.97 USD
14205-834 14206-438 14208-334 14209-020
SP Wilmad-LabGlass Specific Gravity/Pycnometer Bottle Apparatus, SP Industries
Specific Gravity Bottles
With a conical flask body to give maximum stability, these specific gravity bottles/pycnometers come complete with flask, thermometer, and cap.
The 50 and 100 mL sizes are used to obtain the specific gravity of pigments (ASTM Method of Test D153)
Reference No. ASTM Method of Test D 153.
The cap, used for all bottle sizes, has a Standard Taper 5/12 joint.