SP Wilmad-LabGlass Soxhlet Extraction Apparatus, Extra-Large, SP Industries
Supplier: SP Industries
LG-6910-100
14205-526EA 849.43 USD
14205-526 14205-964 14206-012 14207-560 14207-856
SP Wilmad-LabGlass Soxhlet Extraction Apparatus, Extra-Large, SP Industries
Extractors Soxhlet and Soxhlet-Type Extractors
This borosilicate glass X-Large Soxhlet extraction apparatus is used for the continuous extraction of solids with a suitable solvent.
Complete units are supplied with a round bottom boiling flask (14211-116 or 14209-000), a Soxhlet extraction tube (70mm or 100mm approx. I.D.) and an Allihn condenser. The extractor capacity is the solvent volume held in the extractor to the top of the siphon tube. The extractor has a standard taper outer joint at the top for connection to the condenser and a standard taper inner joint at the bottom for connection to the flask.
Ordering information: Soxhlet extraction apparatus uses cellulose, glass microfiber or fritted glass extraction thimbles (not supplied).