SP Wilmad-LabGlass Gas Washing Bottles with Fritted Discs and [ST] 40/35 Stopper Assembly, SP Industries
Supplier: SP Industries
LG-3765-130
14202-894EA 123.3 USD
14202-894 14204-150 14204-854 14205-786 14207-580 14208-256 14208-996 14209-050 14209-668 14210-342 14210-572
SP Wilmad-LabGlass Gas Washing Bottles with Fritted Discs and [ST] 40/35 Stopper Assembly, SP Industries
Bottles Gas Washing Bottles
These borosilicate glass gas washing bottles are generally used to saturate or dry gas streams.
Complete units consist of a bottle and a standard taper 40/35 stopper assembly with an inlet tube that is sealed through the center of a coarse (C) fritted glass disc.
Inlet tube extends nearly to the base of the bottle.