SP Wilmad-LabGlass Siphon ASTM Extraction Apparatus, SP Industries
Supplier: SP Industries
LG-6914-102
14202-994EA 164.56 USD
14202-994 14207-078
SP Wilmad-LabGlass Siphon ASTM Extraction Apparatus, SP Industries
Extractors Automatic Extractor Systems
A block-tin condenser fits within a 400 ml flask and this borosilicate glass extraction cup with siphon tube is suspended with wire from the condenser.
  • Borosilicate glass
Reference: ASTM D 297 Methods for Rubber Products; ASTM D 305 Test Method for Solvent-Extractable Material in Black Pigments.