SP Wilmad-LabGlass Squibb Separatory Funnels with Glass Stopper and PTFE Plug, Drip Tip, SP Industries
Supplier: SP Industries
LG-8431T-108
14203-486EA 244.97 USD
14203-486 14205-410 14207-178 14208-568 14208-570 14203-264 14206-052 14209-438 14209-872
SP Wilmad-LabGlass Squibb Separatory Funnels with Glass Stopper and PTFE Plug, Drip Tip, SP Industries
Funnels Separating Funnels
Featuring bottom stopcock with PTFE plug and glass stopper, these Squibb separatory funnels have a lower standard taper inner joint with a sealed-in drip tube.