SP Wilmad-LabGlass Distilling Adapter with 75° Angle Side Arm, SP Industries
Supplier: SP Industries
LG-1530-100
14203-512EA 226.55 USD
14203-512
SP Wilmad-LabGlass Distilling Adapter with 75° Angle Side Arm, SP Industries
Distillation Components Distillation Adapters
This borosilicate glass three-way adapter has a Standard Taper 10/30 outer joint on top to accommodate a thermometer, bottom inner Standard Taper 24/40 joint, and 75° angle inner side joint of same size.