SP Wilmad-LabGlass Vacuum Sublimation Apparatus, SP Industries
Supplier: SP Industries
ML-1800-700
14202-640EA 190.62 USD
14202-640 14203-410 14203-452
SP Wilmad-LabGlass Vacuum Sublimation Apparatus, SP Industries
Condensers
This sublimation apparatus is designed for purification by sublimation of quantities of approximately 0.5 to 500mg, under vacuum.
This device consists of a cylindrical, flat bottomed flask for sample and a water-cooled finger type condenser. The condenser features an outer short skirt with a Standard Taper 24/25 outer joint to fit over the neck of the flask. Above the joint there is a side hose connector for attachment to vacuum source.