SP Wilmad-LabGlass Gas Washing Tubes, SP Industries
Supplier: SP Industries
ML-1490-702
14202-916EA 255.67 USD
14202-916 14203-366 14206-998 14210-030
SP Wilmad-LabGlass Gas Washing Tubes, SP Industries
Gas Tubes
These borosilicate glass gas washing tubes come complete with tube and a stopper equipped with Standard Taper joints.
Stopper has two hose connections and a center tube with a fine porosity fritted cylinder at its base.