SP Wilmad-LabGlass Washing Bottles with Fritted Cylinders, SP Industries
Supplier: SP Industries
LG-3762-114
14202-734EA 123.11 USD
14202-734 14202-736 14202-834 14202-836 14202-944 14202-946 14203-560 14203-562 14204-542 14204-544 14204-848 14207-450 14207-452 14209-428 14209-430 14209-906 14209-908
SP Wilmad-LabGlass Washing Bottles with Fritted Cylinders, SP Industries
Bottles Gas Washing Bottles
These gas washing bottles are made of borosilicate glass and have a stopper component that contains gas inlet and outlet tubes.
The lower inlet tube, extending nearly to the base of the bottle, contains a Coarse or Extra-Coarse sealed-on fritted cylindrical disc (12 mm Ext.Ø×25 mm length), providing a stream of fine bubbles into the bottle.
Bottle and stopper are joined by a Standard Taper 29/42 joint.
Delivery information: Complete units are supplied with a plastic clamp (LG-1004-110).