SP Wilmad-LabGlass Hubbard Specific Gravity Bottle, SP Industries
Supplier: SP Industries
LG-3541-100
14202-630EA 298.71 USD
14202-630
SP Wilmad-LabGlass Hubbard Specific Gravity Bottle, SP Industries
Specific Gravity Bottles
This borosilicate glass specific gravity bottle is used in standard test methods for specific gravity of a variety of semi-solid materials and emulsions, such as asphalt, cements, road tars, road oils, and soft tar pitches
The solid Standard Taper stopper has a hole approximately 1.6mm in diameter through the center to allow excess sample to escape.
Certifications: Reference: ASTM D 70, AASHO Method T 43.