SP Wilmad-LabGlass Distilling Column/Condenser, SP Industries
Supplier: SP Industries
LG-5794-106
14202-528EA 133.86 USD
14202-528 14202-530 14203-740 14209-966 14209-968
SP Wilmad-LabGlass Distilling Column/Condenser, SP Industries
Distillation Components Distillation Columns
These distilling columns come with indentations for random packing support, and water jacket that allows for alternative use as a condenser.
  • Made of borosilicate glass
They have an outer joint at top and a Standard Taper inner drip tip at bottom.