SP Wilmad-LabGlass Glass Rod Stirrer Shafts, SP Industries
Supplier: SP Industries
LG-9500-100
14202-140EA 75.9 USD
14202-140 14202-396 14202-444 14206-390 14209-834
SP Wilmad-LabGlass Glass Rod Stirrer Shafts, SP Industries
Stirrers Stirring Shafts
Made of borosilicate glass, these precision ground stirrer shafts are equipped with a side button for attachment of stirrer blades.
NOTE: Use blades LG-9505 through LG-9511T for LG-9500 shafts, ML-1770T blade for ML-1760 shaft, and LG-9510T-100 blade for ML-1765 shaft.