SP Wilmad-LabGlass Fat Extraction Flasks, SP Industries
Supplier: SP Industries
LG-6912-100
14202-366EA 237.94 USD
14202-366 14207-902
SP Wilmad-LabGlass Fat Extraction Flasks, SP Industries
Extractors Soxhlet and Soxhlet-Type Extractors
These borosilicate glass extraction apparatus are used to measure the fat content in dairy and other food products.
  • Reference AOAC 989.05 for fat in milk
  • Top is tooled to accept rubber stoppers (not included)