SP Wilmad-LabGlass Reaction Kettle Heating Mantles, SP Industries
Supplier: SP Industries
LG-8881-110
14200-650EA 746.14 USD
14200-650 14200-916 14201-048 14201-112 14201-138
SP Wilmad-LabGlass Reaction Kettle Heating Mantles, SP Industries
Heaters Heating Mantles
These all-glass constructed heating mantles are for use with LG-8071 through LG-8089 reaction kettles. They provide even heat distribution.
  • Heating circuits have two short lead-in wires connected by a Hubbell plug to a 4' long, 2-wire type SJT cord
  • Depth given is the interior measurement
  • Wattage requirements are for 115 V circuits