SP Wilmad-LabGlass Stainless Steel Quick Release Clamps, SP Industries
Supplier: SP Industries
LG-4632-106
14200-780EA 187.89 USD
14200-780 14200-792 14200-814 14200-862 14200-944
SP Wilmad-LabGlass Stainless Steel Quick Release Clamps, SP Industries
Clamps General Purpose Clamps
Made entirely from stainless steel, these clamps firmly hold reaction vessels with Duran® flanges and lids together
Quick release mechanism has a screw adjustment for tension control of flange.