SP Wilmad-LabGlass Reaction Kettle Heating Mantles, SP Industries
Supplier: SP Industries
LG-8883-110
14200-550EA 908.03 USD
14200-550 14200-714 14200-728 14200-764 14200-990
SP Wilmad-LabGlass Reaction Kettle Heating Mantles, SP Industries
Heaters Heating Mantles
Designed for use with cylindrical reaction kettles LG-8071 through LG-8082, these heating mantles have an aluminum housing exterior.
  • Includes detachable 4 foot, 3-wire cord and locking connector
They are equipped with a 3-wire terminal with one grounded to housing.