SP Wilmad-LabGlass Volatile Oil Apparatus for Oils Lighter than Water in Drugs, SP Industries
Supplier: SP Industries
LG-11125-100
14200-672EA 582.65 USD
14200-672 14201-198 14212-218
SP Wilmad-LabGlass Volatile Oil Apparatus for Oils Lighter than Water in Drugs, SP Industries
Distillation Components Distillation Sets
This distilling apparatus is used to determine volatile oil in drugs by distillation of a weighed quantity of drug in contact with water.
  • The Clevenger type receiver trap included in this apparatus is for use when the oils are lighter than water
  • The condenser rests on top of the receiver
  • There is a discharge stopcock below the return tube
  • Flask and receiver are connected by a Standard Taper 24/40 joint
Made of borosilicate glass, the complete apparatus includes a 300mL round bottom flask (LG-7221), a finger type condenser, and a trap/receiver.
The receiver, graduated to 5mL with 0.1mL increments, has a tube connecting the lower part below the scale with the top of the vapor tube to return excess water to the flask.