Anti-CD90.2 Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: 53-2.1]

Supplier: Biolegend
140319
140319-BLEA 200.6 USD
140319-BL
Anti-CD90.2 Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: 53-2.1]
Antibodies
Brilliant Violet 510™ anti-mouse CD90.2 (Thy1.2) [53-2.1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Does not react with Thy-1.1 (CD90.1); Apps: FC; Size: 125 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD90.2
Clonality: Monoclonal
Clone: 53-2.1
Conjugation: Brilliant Violet® 510
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse;Does not react with Thy-1.1 (CD90.1)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR