SP Wilmad-LabGlass Stainless Steel Stirrer Shafts, SP Industries
Supplier: SP Industries
LG-9502-100
14009-480EA 319.7 USD
14009-480 14009-482 14009-484
SP Wilmad-LabGlass Stainless Steel Stirrer Shafts, SP Industries
Stirrers Stirring Shafts
Precision ground, polished stainless steel stirrer shafts have a 10 mm diameter and are used with bearings with PTFE surface (LG-9475T), or other bearings with a PTFE interface and similar bore.
They are equipped with a side button at bottom for attachment of stirrer blades (LG-9505 through LG-9511T).