SP Wilmad-LabGlass Separable Distilling Heads, SP Industries
Supplier: SP Industries
LG-6290-100
14008-524EA 1530.26 USD
14008-524 14008-526 14008-528 14008-530 14008-532
SP Wilmad-LabGlass Separable Distilling Heads, SP Industries
Distillation Components Distillation Heads
These distilling heads feature a removable cold finger condenser with a Standard Taper 29/42 inner joint.
  • Made of borosilicate glass
Distilling heads have an upper Standard Taper 29/42 outer joint, a lower Standard Taper inner joint, and two stopcocks which are in the pressure equalizing line allowing receiver changes without interrupting the distillation. Thermometer joint is Standard Taper 10/30.