SP Wilmad-LabGlass Ross Miles Foam Test Apparatus, SP Industries
Supplier: SP Industries
LG-3941-100
14007-876EA 1891 USD
14007-876 14007-878 14007-880
SP Wilmad-LabGlass Ross Miles Foam Test Apparatus, SP Industries
Petroleum Testing Apparatus
Designed for testing foaming properties of surface active agents.
  • Reference: ASTM D 1173
Foam test apparatus consists of a 200mL foam pipette with a 2mm PTFE straight bore stopcock, and a foam receiver with graduations at 50mL and 250mL above a 6mm PTFE straight bore stopcock.