SP Wilmad-LabGlass PTFE Joint Sleeves, SP Industries
Supplier: SP Industries
LG-1038T-118
14007-458EA 18.25 USD
14007-458 14007-462 14007-466 14007-468 14007-470 14007-472 14007-474 14007-476 14007-480 14007-484 14007-488
SP Wilmad-LabGlass PTFE Joint Sleeves, SP Industries
NS Joint Sleeves and Connectors Joint Sleeves
Made of PTFE with a nominal thickness of 0.003", these sleeves are fitted onto Standard Taper inner joints to prevent 'freezing' of assembled joints.