VWR® Advanced Multi-Tube Vortexer, 230 V (Export Only)
Supplier: VWR International
14005-832
14005-832EA 5870.2 USD
14005-832
VWR® Advanced Multi-Tube Vortexer, 230 V (Export Only)
Shakers and Mixers Vortex Mixers
Advanced multi-tube vortexer features touchpad controls with independent LED displays for speed and time.
  • 230 V model
  • Process up to 50 samples at a time
  • LED displays for speed and time
  • Pulsing mode
  • Best-in-Class, 5-year warranty
Timer will display elapsed time or, when programmed to user defined limits, will shut off when the time reaches zero. Display will show last used settings even after power has been turned off.
Certifications: RoHS certified. 230 V units are CE Marked.
Ordering information: Unit includes a 92" (234 cm) Euro type plug. Unit is also supplied with a Tray Pad Set for support and one 12 mm Test Tube Foam Rack.
Units with NIST Traceable Certificate provided by an ISO/IEC 17025 accredited laboratory. Multiple data points within the speed range and time function are reported on the certificate with the associated uncertainties.
Caution: 230 V model.