STERI-SILICON®, Veltek
Supplier: Veltek Associates
SSIL-02
14003-392CS 971.68 USD
14003-392
STERI-SILICON®, Veltek
Lubricants
This lubricant and releasing spray is designed to speed processing in heat sealing, packaging, and process machinery
Colorless, inert, and stable from -40 to 260˚C (-40 to 500˚F). Aerosol spray. Filtered at 0.2µm. Gamma-irradiated, double-bagged, and delivered with lot-specific sterile documentation