DEC-GLASS® Glass Cleaner, Veltek
Supplier: Veltek Associates
DG-03-16Z
14003-296CS 377.57 USD
14003-296
DEC-GLASS® Glass Cleaner, Veltek
Cleaning Agents Detergents and Surface Cleansers
DEC-GLASS® was developed for use in cleaning rotation cycles to remove residue left behind from disinfecting agents on glass or plexiglas surfaces.
  • Sterile and formulated with water for injection
  • Designed for use pharmaceutical and biotechnology operations