Orion™ Conductivity Standards, Thermo Scientific
Supplier: Thermo Fisher Scientific
These ready-to-use standards allow five conductivity levels to be easily measured in the field
Standards come in convenient 15mL pouches and certified against standards whose calibrations are traceable to the National Institute of Standards and Technology.
Ordering information: Store at room temperature.