KIMBLE® KIMAX® Heavy Duty Beaker, Low Form, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
14005-4000
13913-527CS 481.5 USD
13913-527
KIMBLE® KIMAX® Heavy Duty Beaker, Low Form, DWK Life Sciences
Beakers
Manufactured from borosilicate glass 3.3.
  • Thickest uniform walls are designed specifically to prevent against breakage
  • Durable, matte marking area for labeling
  • With white graduations
Complies with ASTM Specification E960, Type II requirements.
Note: Not recommended for use on hotplates where significant thermal gradients exist due to lack of agitation of contents or absence of a temperature control.
Order Now
SPECIFICATIONS
Environmentally Preferable