Nunc-Immuno™ BreakApart™ and LockWell™ Breakable Modules, Thermo Scientific
Supplier: Thermo Fisher Scientific
446490 446442 446470 446469 473539
73520-834CS 589.12 USD
73520-834 13586-224 73520-754 13586-222 62409-292
Nunc-Immuno™ BreakApart™ and LockWell™ Breakable Modules, Thermo Scientific
Microplates Immunology Microplates and Strips
Provide maximum flexibility in assaying samples
Modules are made of virgin polystyrene and are available in four surface treatments. The mixed hydrophilic/hydrophobic MaxiSorp™ surface binds to a wide variety of biomolecules, but has been optomized to bind IgG. Recommended for assays where glycoproteins, such as antibodies, are adsorbed into the plastic. The hydrophobic PolySorp™ surface is recommended for the immobilization of hydrophobic biomolecules, such as lipids and hydrophobic proteins. PolySorp™ is less polar and can be used for viral membrane antigens.
The MediSorp™ surface is recommended for adsorption of medium to large biomolecules with hydrophilic and hydrophobic properties, such as immunoglobulins, peptides, and albumins. Non-specific binding is reduced for samples that contain serum or plasma.
The MultiSorp™ surface is recommended for adsorption of glycoproteins, polar lipids, and phospholipids.
MaxiSorp™ and MediSorp™ plates are certified for reproducible binding of IgG. Certified plates guarantee uniform binding capacity of CV <5%, intra well O.D. within ±10% from the mean of the plate. Background reading for all wells within ±0.005 adsorbance units from mean.
Modules permit assaying a whole plate, one or more strips, or individual wells. Both plates are 86x128mm with a 96 MicroWell™ design. BreakApart™ modules consist of a frame with 12 well carriers, each holding a break-apart strip of eight wells. The carriers hold the wells securely and enable easy handling of wells within or outside of the frames. The LockWell™ modules use a locking well frame configuration that keeps the wells in the same horizontal position, even during machine operations. Modules feature C-bottom wells with a total volume of 350µL. The wells have rounded sides for thorough washing and a flat bottom for optical clarity. The U8 Breakable 1x8 Modules are designed for the adsorption of hydrophobic molecules. These polystyrene modules offer high optical quality and a working volume of 250µL.