Concentric Ring Round Water Bath, Boekel Scientific
Supplier: Boekel
14458
13540-044EA 739.63 USD
13540-044
Concentric Ring Round Water Bath, Boekel Scientific
Baths Water Baths, Unstirred
The Round Water Bath is made from brushed stainless steel for corrosion resistance and is supplied with seven concentric rings
The Boekel Scientific Concentric Ring Water Bath accommodates many vessel and tube sizes with the smallest opening of 25mm and largest opening of 203mm.
This water bath provides gentle heating of flasks and bottles while minimizing water loss through evaporation.
Ordering information: This unit includes 7 concentric rings.