VWR® Signature™ High-Speed Microplate Shaker
Supplier: VWR International
80862909 80862910
13500-890EA 9125.41 USD
13500-890 10027-220
VWR® Signature™ High-Speed Microplate Shaker
Shakers and Mixers Microplate Shakers
Ideal for molecular biology applications, mechanical and chemical cell lysis, mixing tissue samples, mixing cytogenetic suspensions, and vortexing cell suspensions.
  • Holds up to 48 microplates
  • Designed to Shake Microplates Up to 12.7 cm (5") High
  • Programmable Digital Speed Control from 600 to 2500 rpm
  • Programmable Digital Timer from 1 to 9999 Seconds (166 Minutes)
  • Programmable “Pulsing” Feature to Enhance Mixing Action
  • Now with Best-in-Class, Exclusive 5-Year Warranty
Shaker is also ideal for emulsification of bipolar viscous fluids and for mixing fluids in small diameter tubes
Tray can be modified for other vessels up to 12.7 cm (5") high and 3.2kg (7lbs.) in weight. Tray assembly height is adjustable from 3.2mm (1/8") to 12.7 cm (5") to accommodate most microplates, especially deep-well microplates. The unit’s 12 suction cup feet and 22.7kg (50lbs.) weight prevent shaker from “walking” across the lab bench.
Pulsing feature allows the user to program the unit to stop and start at a wide range of intervals. The repetitive stopping and starting action enhances the mixing action and is needed for difficult mixing applications, such as emulsification of highly viscous liquids or bipolar liquids. Shaker comes with an RS-232 interface.
Specifically designed to shake and/or vortex microplates in timed or continuous modes. Shaker features programmable speed from 600 to 2500rpm (±25rpm), and programmable timed mode from 1 to 9999 seconds (166 minutes). Speed and time are easily adjusted with the up/down arrows on the front panel. Length of time is displayed for continuous mode, and time remaining is displayed for timed mode.
Certifications: RoHS certified. UL listed. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Contact your Avantor sales representative for more information.