Nalgene® Crimp Finish Serum Vials, PETG, Non-Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
322030-0010
13500-118CS 711.25 USD
13500-118
Nalgene® Crimp Finish Serum Vials, PETG, Non-Sterile, Thermo Scientific
Vials
Nalgene non-sterile serum vials are made for use with stoppers and caps (not included).
  • Non-cytotoxic
  • Non-pyrogenic
  • Packaged in tray less shrink-wrap modules
These serum vials are lighter in weight than glass, and protect against leakage, breakage, and contamination.
Int. Ø: 13 mm