DuPont™ Tychem® 4000 Bib Aprons
Supplier: DuPont
SL274BWH00005000
13452-842CS 387.78 USD
13452-842
DuPont™ Tychem® 4000 Bib Aprons
Disposable Garments
DuPont™ Tychem® 4000 delivers effective protection against a range of chemical environments.
  • Neck loop
  • Waist ties
  • Bound seams
  • One size fits most
Tychem® 4000, utilizes Saranex™ 23-P film laminated to Tyvek® fabric to provide a rugged and durable fabric that resists abrasion. Tychem® 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications include chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.
These bib aprons provide enhanced frontal protection. Bound seams and ties at waist allow for one size to fit most.
Certifications: Thickness meets ASTM D1777.