DuPont™ ProShield® 60 Bib Aprons
Supplier: DuPont
NG273BWH00010000
13451-080CS 214.15 USD
13451-080
DuPont™ ProShield® 60 Bib Aprons
Disposable Garments
ProShield® 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
  • Neck loop
  • Waist ties
  • Bound seams
  • One size fit most
ProShield® 60 garments are made with a microporous film that is laminated to a spunbonded polypropylene substrate. Applications include automotive refinishing, waste cleanup and remediation.
These bib aprons provide enhanced frontal protection. Bound seams around neck and ties at waist allow for one size to fit most.
Certifications: Thickness meets ASTM D751.