CR Bench Mats, Dycem
Supplier: Dycem
CC02/BM/2440
13450-268EA 3704.64 USD
13450-268
CR Bench Mats, Dycem
Surface Protectors Bench Protectors
Designed to safely drain static discharge (108 ohms resistance), this closed cell mat is nonabrasive, nonparticulating, and requires no adhesives or outgassing
The bench mat improves accuracy, precision, and assembly time. Supplied with earthing studs. Color: gray.
Ordering information: Also available in black; contact your Avantor sales representative for more information.